Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RKTVLSKAI EKIKCLFCRI GGSRRERSGQ HCSDSQRTSS AMSGHSRSFI SRELKEISST SQTLLPDLSL PDLSENGLGG RNLLPGVPGM GLAQEDTTSL RTLRISETSD SSQGQDSESV LLVDEAGGSG RAGPAPKGSS LQVTFPSETL NLSEKCI (333-488 aa encoded by BC101534 )
The protein encoded by this gene is a member of the G-protein coupled receptor family. This protein is one of four receptors identified for prostaglandin E2. This receptor can activate T-cell factor signaling. It has been shown to mediate PGE2 induced expression of early growth response 1, regulate the level and stability of cyclooxygenase-2 mRNA, and lead to the phosphorylation of glycogen synthase kinase-3. Knockout studies in mice suggest that this receptor may be involved in the neonatal adaptation of circulatory system, osteoporosis, as well as initiation of skin immune responses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EP1; MGC126583; PGE receptor EP4 subtype; PGE receptor, EP4 subtype; PGE2 receptor EP4 subtype; prostaglandin E receptor 4 (subtype EP4); Prostaglandin E2 receptor EP4 subtype; prostaglandin E2 receptor type 4; Prostanoid EP4 receptor
Gene Aliases: EP4; EP4R; Ptger; PTGER2; PTGER4; Ptgerep4
UniProt ID: (Human) P35408, (Mouse) P32240, (Rat) P43114
Entrez Gene ID: (Human) 5734, (Mouse) 19219, (Rat) 84023
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support