Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVRTKTWTL KKHFVGYPTN SDFELKTSEL PPLKNGEVLL EALFLTVDPY MRVAAKRLKE GDTMMGQQVA KVVESKNVAL PKGTIVLASP GWTTHSISDG KDLEKLLTEW PDTIPLSLAL GTVGMPGLTA YFGLLEICGV KGGETVMVNA AAGAVGSVVG QIAKLKGCKV VGAVGSDEKV AYLQKLGFDV VFNYKTVESL EETLKKASPD GYDCYFDNVG GEFSNTVIGQ MKKFGRIAIC GAISTYNRTG PLPPGPPPEI VIYQELRMEA FVVYRWQGDA RQKALKDLLK WVLEGKIQYK EYIIEGFENM PAAFMGMLKG DNLGKTIVKA (1-329 aa encoded by BC035228)
PTGR1 encodes an enzyme that is involved in the inactivation of the chemotactic factor, leukotriene B4. The encoded protein specifically catalyzes the NADP+ dependent conversion of leukotriene B4 to 12-oxo-leukotriene B4. A pseudogene of this gene is found on chromosome 1. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 15-oxoprostaglandin 13-reductase; D3T-inducible gene 1 protein; DIG-1; Dithiolethione-inducible gene 1 protein; dithiolethione-inducible gene-1; FLJ99229; Leukotriene B4 12-hydroxydehydrogenase; MGC34943; NAD(P)H-dependent alkenal/one oxidoreductase; NADP-dependent leukotriene B4 12-hydroxydehydrogenase; PRG-1; Prostaglandin reductase 1; zinc binding alcohol dehydrogenase domain containing 3
Gene Aliases: 2510002C21Rik; Dig1; LTB4DH; PGR1; PTGR1; ZADH3
UniProt ID: (Human) Q14914, (Mouse) Q91YR9, (Rat) P97584
Entrez Gene ID: (Human) 22949, (Mouse) 67103, (Rat) 192227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support