Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: SDCGEWQWSV CVPTSGDCGL GTREGTRTGA ECKQTMKTQR CKIPCNWKKQ FGAECKYQFQ AWGECDLNTA LKTRTGSLKR ALHNAECQKT VTISKPCGKL TKPKPQAESK
Pleiotrophin (PTN) is an 18 kDa secreted heparin and chondroitin-binding protein that is a regulatory cytokine critical to the early development of blood vessels and neurons. High PTN serum levels are associated with a variety of solid tumors. It was shown that patients with multiple myeloma (MM) also have elevated serum levels of this protein. The expression of this protein is very limited in normal adult tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HB-GAM; HBBM; HBGF-8; HBNF; HBNF-1; heparin affin regulatory protein; Heparin-binding brain mitogen; Heparin-binding growth factor 8; Heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; OSF-1; Osteoblast-specific factor 1; Pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); PTN
Gene Aliases: HARP; HB-GAM; HBBM; HBGF-8; HBGF8; HBNF; HBNF-1; HBNF1; NEGF1; OSF-1; PTN
UniProt ID: (Human) P21246
Entrez Gene ID: (Human) 5764
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support