Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MARMNRPAPV EVTYKNMRFL ITHNPTNATL NKFIEELKKY GVTTIVRVCE ATYDTTLVEK EGIHVLDWPF DDGAPPSNQI VDDWLSLVKI KFREEPGCCI AVHCVAGLGR APVLVALALI EGGMKYEDAV QFIRQKRRGA FNSKQLLYLE KYRPKMRLRF KDSNGHRNNC CIQ
The protein encoded by this gene belongs to a small class of prenylated protein tyrosine phosphatases (PTPs), which contains a PTP domain and a characteristic C-terminal prenylation motif. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. This tyrosine phosphatase is a nuclear protein, but may primarily associate with plasma membrane. The surface membrane association of this protein depends on its C-terminal prenylation. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which implicated its role in the tumorigenesis. Studies in rat suggested that this gene may be an immediate-early gene in mitogen-stimulated cells. PRL phosphatases increase cell proliferation by stimulating progression from G1 into S phase. PRL-1 function is regulated in a cell cycle-dependent manner and implicate PRL-1 in regulating progression through mitosis, possibly by modulating spindle dynamics.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp779M0721; PRL-1; protein tyrosine phosphatase 4a1; Protein tyrosine phosphatase type IVA 1; protein tyrosine phosphatase type IVA protein 1; Protein-tyrosine phosphatase 4a1; Protein-tyrosine phosphatase of regenerating liver 1; PTP(CAAXI); PTP4A1 protein; PTP4A2; RP5-1148A21.2
Gene Aliases: AA415290; AU019864; C130021B01; HH72; PRL-1; PRL1; PTP(CAAX1); PTP4A1; PTPCAAX1
UniProt ID: (Human) Q93096, (Rat) Q78EG7, (Mouse) Q63739
Entrez Gene ID: (Human) 7803, (Rat) 29463, (Mouse) 19243
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support