Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MPFGLKLRRT RRYNVLSKNC FVTRIRLLDS NVIECTLSVE STGQECLEAV AQRLELRETH YFGLWFLSKS QQARWVELEK PLKKHLDKFA NEPLLFFGVM FYVPNVSWLQ QEATRYQYYL QVKKDVLEGR LRCTLDQVIR LAGLAVQADF GDYNQFDSQD FLREYVLFPM DLALEEAVLE ELTQKVAQEH KAHSGILPAE AELMYINEVE RLDGFGQEIF PVKDNHGNCV HLGIFFMGIF VRNRIGRQAV IYRWNDMGNI THNKSTILVE LINKEETALF HTDDIENAKY ISRLFATRHK FYKQNKICTE QSNSPPPIRR QPTWSRSSLP RQQPYILPPV HVQCGEHYSE T
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal noncatalytic domain similar to that of band 4.1 superfamily cytoskeleton-associated proteins, which suggested the membrane or cytoskeleton localization of this protein. The specific function of this PTP has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoskeletal-associated protein tyrosine phosphatase; Protein-tyrosine phosphatase pez; Tyrosine-protein phosphatase non-receptor type 14
Gene Aliases: PEZ; PTP36; PTPD2; PTPN14
UniProt ID: (Human) Q15678
Entrez Gene ID: (Human) 5784
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support