Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGASFWPIR QAREQQRRAL SFRQTSWLSE PPLGPAPHLS MVQAHGGRSR AQPLTLSLGA AMTQPPPEKT PAKKHVRLQE RRGSNVALML DVRSLGAVEP ICSVNTPREV TLHFLRTAGH PLTRWALQRQ PPSPKQLEEE FLKIPSNFVS PEDLDIPGHA SKDRYKTILP NPQSRVCLGR AQSQEDGDYI NANYIRGYDG KEKVYIATQG PMPNTVSDFW EMVWQEEVSL IVMLTQLREG KEKCVHYWPT EEETYGPFQI RIQDMKECPE YTVRQLTIQY QEERRSVKHI LFSAWPDHQT PESAGPLLRL VAEVEESPET AAHPGPIVVH CSAGIGRTGC FIATRIGCQQ LKARGEVDIL GIVCQLRLDR GGMIQTAEQY QFLHHTLALY AGQLPEEPSP (1-399 aa encoded by BC001746)
The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This gene is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The noncatalytic N-terminus of this PTP can interact with MAP kinases and suppress the MAP kinase activities. This PTP was shown to be involved in the regulation of T cell antigen receptor signaling, which was thought to function through dephosphorylating the molecules related to MAP kinase pathway. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dual specificity phosphatase 1; Hematopoietic protein-tyrosine phosphatase; HEPTP; Protein-tyrosine phosphatase LC-PTP; protein-tyrosine phosphatase, non-receptor type 7; protein-tyrosine phosphatase, nonreceptor-type, stress induced; Tyrosine-protein phosphatase non-receptor type 7
Gene Aliases: BPTP-4; C920001D21Rik; HEPTP; LC-PTP; Lcptp; LPTP; PTPN7; PTPNI
UniProt ID: (Human) P35236, (Rat) P49445, (Mouse) Q8BUM3
Entrez Gene ID: (Human) 5778, (Rat) 246781, (Mouse) 320139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support