Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MGHLPTGIHG ARRLLPLLWL FVLFKNATAF HVTVQDDNNI VVSLEASDVI SPASVYVVKI TGESKNYFFE FEEFNSTLPP PVIFKASYHG LYYIITLVVV NGNVVTKPSR SITVLTKPLP VTSVSIYDYK PSPETGVLFE IHYPEKYNVF TRVNISYWEG KDFRTMLYKD FFKGKTVFNH WLPGMCYSNI TFQLVSEATF NKSTLVEYSG VSHEPKQHRT APYPPQNISV RIVNLNKNNW EEQSGNFPEE SFMRSQDTIG KEKLFHFTEE TPEIPSGNIS SGWPDFNSSD YETTSQPYWW DSASAAPESE DEFVSVLPME YENNSTLSET EKSTSGSFSF FPVQMILTWL PPKPPTAFDG FHIHIEREEN FTEYLMVDEE AHEFVAELKE PGKYKLSVTT FSSSGSCETR KSQSAKSLSF YISPSGEWIE ELTEKPQHVS VHVLSSTTAL MSWTSSQENY NSTIVSVVSL TCQKQKESQR LEKQYCTQVN SSKPIIENLV PGAQYQVVIY LRKGPLIGPP SDPVTFAIVP TGIKDLMLYP LGPTAVVLSW TRPYLGVFRK YVVEMFYFNP ATMTSEWTTY YEIAATVSLT ASVVIFP
Glomerular epithelial protein 1 (GLEPP1), also designated protein tyrosine phosphatase U2 (PTPase U2) or receptor-type tyrosine-protein phosphatase O (PTPRO), belongs to the protein-tyrosine phosphatase family. GLEPP1 is a Type I membrane protein containing 8 fibronectin type-III domains and 1 tyrosine-protein phosphatase domain. GLEPP1 is expressed primarily in the glomeruli of the kidney, but is also detected in placenta, lung and brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FMI; Glomerular epithelial protein 1; hPTP-J; osteoclastic transmembrane protein-tyrosine phosphatase; PCP2; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; Protein tyrosine phosphatase U2; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP-U2; PTPase U2; PTPRU; R-PTP-O; Receptor-type tyrosine-protein phosphatase O
Gene Aliases: GLEPP1; NPHS6; PTP-OC; PTP-U2; PTPRO; PTPROT; PTPU2; R-PTP-O
UniProt ID: (Human) Q16827
Entrez Gene ID: (Human) 5800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support