Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: colon or skeletal muscle. Human brain pancreas
Cellular Location: Cell junction. Cell membrane. Single-pass type I membrane protein.
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP possesses an extracellular region, a single transmembrane region, and two tandem intracellular catalytic domains, and thus represents a receptor-type PTP. The extracellular region contains a meprin-A5 antigen-PTP (MAM) domain, Ig-like and fibronectin type III-like repeats. This PTP was thought to play roles in cell-cell recognition and adhesion. Studies of the similar gene in mice suggested the role of this PTP in early neural development. The expression of this gene was reported to be regulated by phorbol myristate acetate (PMA) or calcium ionophore in Jurkat T lymphoma cells. Alternatively spliced transcript variants have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GLEPP1; Pancreatic carcinoma phosphatase 2; PCP-2; pi R-PTP-Psi; protein tyrosine phosphatase, receptor type, U; Protein-tyrosine phosphatase J; Protein-tyrosine phosphatase pi; Protein-tyrosine phosphatase receptor omicron; PTP pi; PTP-J; PTP-RO; R-PTP-psi; R-PTP-U; Receptor protein tyrosine phosphatase hPTP-J; Receptor-type protein-tyrosine phosphatase psi; Receptor-type tyrosine-protein phosphatase U
Gene Aliases: FMI; hPTP-J; PCP-2; PCP2; PTP; PTP-J; PTP-PI; PTP-RO; PTPPSI; PTPRO; PTPRU; PTPU2; R-PTP-PSI; R-PTP-U
UniProt ID: (Human) Q92729
Entrez Gene ID: (Human) 10076
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support