Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPSKSLVME YLAHPSTLGL AVGVACGMCL GWSLRVCFGM LPKSKTSKTH TDTESEASIL GDSGEYKMIL VVRNDLKMGK GKVAAQCSHA AVSAYKQIQR RNPEMLKQWE YCGQPKVVVK APDEETLIAL LAHAKMLGLT VSLIQDAGRT QIAPGSQTVL GIGPGPADLI DKVTGHLKLY (1-180 aa encoded by BC006807)
The protein encoded by this gene is a mitochondrial protein with two putative domains, an N-terminal mitochondrial localization sequence, and a UPF0099 domain. In vitro assays suggest that this protein possesses peptidyl-tRNA hydrolase activity, to release the peptidyl moiety from tRNA, thereby preventing the accumulation of dissociated peptidyl-tRNA that could reduce the efficiency of translation. This protein also plays a role regulating cell survival and death. It promotes survival as part of an integrin-signaling pathway for cells attached to the extracellular matrix characterized by intellectual disability, postnatal microcephaly, progressive cerebellar atrophy, hearing impairment, polyneuropathy, failure to thrive, and organ fibrosis with exocrine pancreas insufficiency. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bcl-2 inhibitor of transcription; Bcl-2 inhibitor of transcription 1; cilia and flagella associated protein 37; OTTHUMP00000205936; Peptidyl-tRNA hydrolase 2, mitochondrial; PTH 2
Gene Aliases: A230072I16Rik; BIT1; CFAP37; CGI-147; IMNEPD; PTH; PTH 2; PTH2; PTRH2; RGD1306819
UniProt ID: (Human) Q9Y3E5, (Mouse) Q8R2Y8
Entrez Gene ID: (Human) 51651, (Rat) 287593, (Mouse) 217057
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support