Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: SLWVNGELAA TTVEMATGHI VPEGGILQIG QEKNGCCVGG GFDETLAFSG RLTGFNIWDS VLSNEEIRET GGAESCHIRG NIVGWGVTEI QPHGGAQYV
Pentraxins are a protein superfamily that is characterized by a cyclic multimeric structure. Ptx3, also known as tumor necrosis factor-stimulated gene sequence-14 (TSG14), is a secreted pattern-recognition receptor that has a non-redundant role in resistance to selected microbial agents. Ptx3 belongs to the family of "long pentraxins", which have C-terminal pentraxin domains and novel amino-terminal domains. Ptx3 binds selected pathogens, including Aspergillus fumigatus, Pseudomonas aeruginosa and Salmonella typhimurium. It is synthesized in IgA glomerulonephritis and activates mesangial cells. Secretion of Ptx3 in adipose cells can be induced by TNF. Ptx3 is also involved in amplification of inflammatory reactions and regulation of innate immunity. The human PTX3 gene maps to chromosome 3q25.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: long pentraxin 3; pentaxin related; Pentaxin-related protein PTX3; pentraxin 3; pentraxin related; Pentraxin-related protein PTX3; TNF alpha-induced protein 5; TSG-14; Tumor necrosis factor alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14
Gene Aliases: AI607804; PTX3; TNFAIP5; TSG-14; TSG14
UniProt ID: (Human) P26022, (Mouse) P48759
Entrez Gene ID: (Human) 5806, (Mouse) 19288, (Rat) 689388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support