Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
This product is predicted to work in Immunoprecipitation at a dilution of 1:10-1:50.
PUM1 (pumilio homolog 1) is a sequence-specific RNA-binding protein that acts as a post-transcriptional repressor by binding the 3'-UTR of mRNA targets. It is a member of the PUF family comprised of evolutionarily conserved RNA-binding proteins. PUM1 binds to an RNA consensus sequence, the Pumilio Response Element (PRE), and 5'-UGUANAUA-3' that is related to the Nanos Response Element (NRE). PUM1 mediates post-transcriptonal repression of transcripts via different mechanisms - acts via direct recruitment of the CCR4-POP2-NOT deadenylase leading to translational inhibition and mRNA degradation. Mutations to the gene can result in spinocerebellar ataxia 47.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HsPUM; pumilio 1; Pumilio homolog 1; pumilio RNA-binding family member 1; pumilio-1
Gene Aliases: AA517475; HSPUM; KIAA0099; mKIAA0099; PUM1; PUMH; PUMH1; PUML1; Pumm
UniProt ID: (Human) Q14671, (Mouse) Q80U78
Entrez Gene ID: (Human) 9698, (Mouse) 80912
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support