Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: DIMPSGRSRL LEDFRNNRFP NLQLRDLIGH IVEFSQDQHG SRFIQQKLER ATPAERQMVF NEILQAAYQL MTDVFGNYVI QKFFEFGSLD QKLALATR
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36528; MGC138251; MGC138253; OTTHUMP00000200539; Pumilio homolog 2; pumilio RNA-binding family member 2; Pumilio-2
Gene Aliases: KIAA0235; PUM2; PUMH2; PUML2
UniProt ID: (Human) Q8TB72
Entrez Gene ID: (Human) 23369
Molecular Function: RNA metabolism protein nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support