Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: PC-3, Mouse testis, Rat testis
Immunogen sequence: MNHDFQALAL ESRGMGELLP TKKFWEPDDS TKDGQKGIFL GDDEWRETAW GASHHSMSQP IMVQRRSGQG FHGNSEVNAI LSPRSESGGL GVSMVEYVLS
LZTS2 is a negative regulator of katanin-mediated microtubule severing and release from the centrosome. LZTS2 is required for central spindle formation and the completion of cytokinesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36528; MGC138251; MGC138253; OTTHUMP00000200539; PUM1; pumilio 2; Pumilio homolog 2; pumilio RNA-binding family member 2; Pumilio-2
Gene Aliases: 5730503J23Rik; KIAA0235; mKIAA0235; PUM2; PUMH2; PUML2; Pumm2
UniProt ID: (Human) Q8TB72, (Mouse) Q80U58
Entrez Gene ID: (Human) 23369, (Rat) 298874, (Mouse) 80913
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support