Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MNRSRQVTCV AWVRCGVAKE TPDKVELSKE EVKRLIAEAK EKLQEEGGGS DEEETGSPSE DGMQSARTQA RPREPLEDGD PEDDRTLDDD ELAEYDLDKY DEEGDPDAET LGESLLGLTV YGSNDQDPYV TLKDTEQYER EDFLIKPSDN LIVCGRAEQD QCNLDVHVYN QEEDSFYVHH DILLSAYPLS VEWLNFDPSP DDSTGNYIAV GNMTPVIEVW DLDIVDSLEP VFTLGSKLSK KKKKKGKKSS SAEGHTDAVL DLSWNKLIRN VLASASADNT VILWDMSLGK PAASLAVHTD KVQTLQFHPF EAQTLISGSY DKSVALYDCR SPDESHRMWR FSGQIERVTW NHFSPCHFLA STDDGFVYNL DARSDKPIFT LNAHNDEISG LDLSSQIKGC LVTASADKYV KIWDILGDRP SLVHSRDMKM GVLFCSSCCP DLPFIYALRG QKEGLRVWDI STVSSVNEAF GRRERLVLGS ARNSSISGPF GSRSSDTPME S
The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C20orf152; endonuclein; Keratinocyte protein IEF SSP 9502; nuclear phosphoprotein similar to S. cerevisiae PWP1; Periodic tryptophan protein 1 homolog
Gene Aliases: IEF-SSP-9502; PWP1
UniProt ID: (Human) Q13610
Entrez Gene ID: (Human) 11137
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support