Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: NLKYLYLYKN EIQSIDRQAF KGLASLEQLY LHFNQIETLD PDSFQHLPKL
Sequence homology: Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hsPxd01; Melanoma-associated antigen MG50; p53-responsive gene 2 protein; Peroxidasin 1; Peroxidasin homolog; Vascular peroxidase 1
Gene Aliases: COPOA; D2S448; D2S448E; KIAA0230; MG50; PRG2; PXD01; PXDN; PXN; VPO; VPO1
UniProt ID: (Human) Q92626
Entrez Gene ID: (Human) 7837
Molecular Function:
peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support