Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
MA5-31809 has been tested in indirect ELISA.
This gene product belongs to the nuclear receptor superfamily, members of which are transcription factors characterized by a ligand-binding domain and a DNA-binding domain. The encoded protein is a transcriptional regulator of the cytochrome P450 gene CYP3A4, binding to the response element of the CYP3A4 promoter as a heterodimer with the 9-cis retinoic acid receptor RXR. It is activated by a range of compounds that induce CYP3A4, including dexamethasone and rifampicin. Several alternatively spliced transcripts encoding different isoforms, some of which use non-AUG translation initiation codon, have been described for this gene. Additional transcript variants exist, however, they have not been fully characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group I, member 2; ONR 1; Orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; PAR 1; PAR 2; PAR q; pregnane X nuclear receptor variant 2; Pregnane X receptor; Steroid and xenobiotic receptor; SXR
Gene Aliases: BXR; NR1I2; ONR1; PAR; PAR1; PAR2; PARq; PRR; PXR; SAR; SXR
UniProt ID: (Human) O75469
Entrez Gene ID: (Human) 8856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support