Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAKPLTDQEK RRQISIRGIV GVENVAELKK SFNRHLHFTL VKDRNVATTR DYYFALAHTV RDHLVGRWIR TQQHYYDKCP KRVYYLSLEF YMGRTLQNTM INLGLQNACD EAIYQLGLDI EELEEIEEDA GLGNGGLGRL AACFLDSMAT LGLAAYGYGI RYEYGIFNQK IRDGWQVEEA DDWLRYGNPW EKSRPEFMLP VHFYGKVEHT NTGTKWIDTQ VVLALPYDTP VPGYMNNTVN TMRLWSARAP NDFNLRDFNV GDYIQAVLDR NLAENISRVL YPNDNFFEGK ELRLKQEYFV VAATLQDIIR RFKASKFGST RGAGTVFDAF PDQVAIQLND THPALAIPEL MRIFVDIEKL PWSKAWELTQ KTFAYTNHTV LPEALERWPV DLVEKLLPRH LEIIYEINQK HLDRIVALFP KDVDRLRRMS LIEEEGSKRI NMAHLCIVGS HAVNGVAKIH SDIVKTKVFK DFSELEPDKF QNKTNGITPR RWLLLCNPGL AELIAEKIGE DYVKDLSQLT KLHSFLGDDV FLRELAKVKQ ENKLKFSQFL ETEYKVKINP SSMFDVQVKR IHEYKRQLLN CLHVITMYNR IKKDPKKLFV PRTVIIGGKA APGYHMAKMI IKLITSVADV VNNDPMVGSK LKVIFLENYR VSLAEKVIPA TDLSEQISTA GTEASGTGNM KFMLNGALTI GTMDGANVEM AEEAGEENLF IFGMRIDDVA ALDKKGYEAK EYYEALPELK LVIDQIDNGF FSPKQPDLFK DIINMLFYHD RFKVFADYEA YVKCQDKVSQ LYMNPKAWNT MVLKNIAASG KFSSDRTIKE YAQNIWNVEP SDLKISLSNE SNKVNGN
This gene encodes a homodimeric protein that catalyses the cleavage of alpha-1,4-glucosidic bonds to release glucose-1-phosphate from liver glycogen stores. This protein switches from inactive phosphorylase B to active phosphorylase A by phosphorylation of serine residue 15. Activity of this enzyme is further regulated by multiple allosteric effectors and hormonal controls. Humans have three glycogen phosphorylase genes that encode distinct isozymes that are primarily expressed in liver, brain and muscle, respectively. The liver isozyme serves the glycemic demands of the body in general while the brain and muscle isozymes supply just those tissues. In glycogen storage disease type VI, also known as Hers disease, mutations in liver glycogen phosphorylase inhibit the conversion of glycogen to glucose and results in moderate hypoglycemia, mild ketosis, growth retardation and hepatomegaly. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycogen liver; Glycogen phosphorylase, liver form; GPLL
Gene Aliases: GSD6; PYGL
UniProt ID: (Human) P06737
Entrez Gene ID: (Human) 5836
Molecular Function:
glycosyltransferase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support