Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody is specific to PYGM.
This gene encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycogen muscle; Glycogen phosphorylase, muscle form; GPMM; muscle (McArdle syndrome); muscle glycogen phosphorylase; Myophosphorylase; Phosphorylase glycogen; Phosphorylase, glycogen; muscle (McArdle syndrome)
Gene Aliases: AI115133; Muscpho; PG; PYGM
UniProt ID: (Human) P11217, (Mouse) Q9WUB3
Entrez Gene ID: (Human) 5837, (Rat) 24701, (Mouse) 19309
Molecular Function:
glycosyltransferase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support