Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPAF1; Paf1, RNA polymerase II associated factor, homolog; Pancreatic differentiation protein 2; RNA polymerase II-associated factor 1 homolog
Gene Aliases: 5730511K23Rik; AA675045; F23149_1; PAF1; PD2
UniProt ID: (Human) Q8N7H5, (Mouse) Q8K2T8
Entrez Gene ID: (Human) 54623, (Mouse) 54624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support