Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin's different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 90kDa actin-associated protein palladin; FLJ22190; FLJ38193; FLJ39139; myoneurin; Palladin; palladin isoform; Sarcoma antigen NY-SAR-77; SIH002
Gene Aliases: 2410003B16Rik; 6030492A02; CGI-151; CGI151; KIAA0992; MYN; PALLD; PNCA1; SIH002
UniProt ID: (Human) Q8WX93, (Mouse) Q9ET54
Entrez Gene ID: (Human) 23022, (Mouse) 72333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support