Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive Samples: Mouse intestine, Rat intestine
Immunogen sequence: QYSPNTQQGR TSIVHLFEWR WVDIALECER YLAPKGFGGV QVSPPNENVA IYNPFRPWWE RYQPVSYKLC TRSGNEDEFR NMVTRCNNVG VRIYVDAVIN HMCGNAVSAG TSSTCGSYFN PGSRDFPAVP YSGWDFNDGK CKTGSGDIEN YNDATQVRDC RLTGLLDLAL EKDYVRSKIA EYMNHLIDIG VAGFRLDASK HMWPGDIKAI LDKLHNLNSN WFPAGSKPFI YQEVIDLGGE PIKSSDYFGN GRVTEFKYGA KLGTVIRKWN GEKMSYLKNW GEGWGFVPSD RALVFVDNHD NQRGHGAGGA SILTFWDARL YKMAVGFMLA HPYGFTRVMS SYRWPRQFQN GNDVNDWVGP PNNNGVIKEV TINPDTTCGN DWVCEHRWRQ IRNMVIFRNV VDGQPFTNWY DNGSNQVAFG RGNRGFIVFN NDDWSFSLTL QTGLPAGTYC DVISGDKING NCTGIKIYVS DDGKAHFSIS NSAEDPFIAI HAESKL
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch andglycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; alpha-amylase; Alpha-amylase 2B; amylase; amylase 2-2, pancreatic; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; Carcinoid alpha-amylase; glycogenase; PA; Pancreatic alpha-amylase; Pancreatic alpha-amylase 2a5; pancreatic amylase alpha 2A
Gene Aliases: Amy-X; AMY2; Amy2-1; Amy2-2; AMY2A; Amy2a5; AMY2B; AMY3; ENSMUSG00000074267; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA
UniProt ID: (Human) P04746, (Human) P19961, (Mouse) P00688
Entrez Gene ID: (Human) 279, (Human) 280, (Mouse) 100043207, (Mouse) 100043684, (Mouse) 545562
Molecular Function:
amylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support