Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitution: 1 X 25 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
Pannexins constitute a new family of gap junction type proteins. They are highly conserved in worms, mollusks, insects and mammals and are predicted to have four transmembrane regions, two extracellular loops, one intracellular loop and intracellular N- and C-termini. Besides the connexin proteins, Pannexin 1 and 2 seem to be molecular components of neuronal gap junctions ("electrical synapses") that are thought to form connections between principal cells in the hippocampus. Functional expression in paired Xenopus oocytes indicated that pannexins are capable of forming communicating junctions. In addition to forming gap junction channels in paired oocytes, Pannexin 1 can also form a mechanosensitive and ATP-permeable channel in the nonjunctional plasma membrane. Pannexin 1 and 3 are both glycoproteins with a cell surface distribution profile and life cycle dynamics distinct from connexin 43. Pannexin 3 expression was prevalent in skin and cartilage. In transiently expressing cells both pannexin 1 and 3 were incapable of forming intercellular channels but assembled into functional cell surface channels. Suggested roles in the ischemic death of neurons, schizophrenia, inflammation and tumor suppression have drawn attention for properties and cellular functions of pannexins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gap junction protein pannexin 3; Pannexin-3
Gene Aliases: 3230401P04; 4833413G11Rik; PANX3; PX3
UniProt ID: (Human) Q96QZ0, (Mouse) Q8CEG0, (Rat) P60572
Entrez Gene ID: (Human) 116337, (Mouse) 208098, (Rat) 315567
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support