Invitrogen
The MA1-7316 antibody reacts with Parainfluenza virus type 3 in viral and infected tissue samples.
MA1-7316 has been successfully used in ELISA and Immunofluorescence applications.
The MA1-7316 antibody was raised against Parainfluenza Type 3 antigen.
Protein Aliases: canine parainfluenza virus; hPIV-3; PIV-3