Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MGMCSRQERI QKDIDVVIQK SRAEKDCLFA DFRYSDSTFT FTYVGGPRSV SYSVHVSEDY PDNTYVSSSE NDEDVLVTTE PIPVIFHRIA TELRKTNDIN CCLSIKSKLQ KENGEESRQN STVEEDSEGD NDSEEFYYGG QVNYDGELHK HPQLEADLSA VREIYGPHAV SLREYGAIDD VDIDLHIDVS FLDEEIAVAW EVIRTEPIIV RLHCSLTQYL NGPVPTVDVF QISTKERFGL GHQLKKIMQT FVTQQWKQSK EKSNCLHNKK LSEKKVKSPL HLFSTLRRSP SYPPPGCGKS; Positive Samples: Mouse skeletal muscle, Rat skeletal muscle
Poly(ADP-ribosylation) is a method of DNA damage-dependent posttranslational modification that helps to rescue injured proliferating cells from cell death. The PARP (poly(ADP-ribose) polymerase) proteins comprise a superfamily of enzymes that functionally modify histones and other nuclear proteins, thereby preventing cell death.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 16; ARTD16; PARP-8; poly (ADP-ribose) polymerase family, member 8; Poly [ADP-ribose] polymerase 8; Protein mono-ADP-ribosyltransferase PARP8
Gene Aliases: 2810430O08Rik; ARTD16; D13Ertd275e; PARP8; pART16
UniProt ID: (Human) Q8N3A8
Entrez Gene ID: (Human) 79668, (Mouse) 52552, (Rat) 294762
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support