Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-Kappa-B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon; and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase pellino homolog 1; pellino homolog 1; Pellino-1; Pellino-related intracellular-signaling molecule; pellino1; protein pellino homolog 1; RING-type E3 ubiquitin transferase pellino homolog 1
Gene Aliases: 2810468L03Rik; A930031K15Rik; AA409794; AI586297; D11Ertd676e; PELI1; PRISM
UniProt ID: (Human) Q96FA3, (Mouse) Q8C669
Entrez Gene ID: (Human) 57162, (Mouse) 67245
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support