Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Reconstitution: 1 X 25 µL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
SLC15A2 belongs to the PTR2/POT transporter (TC 2.A.17) family. It is a multi-pass membrane protein. The expression/activity of PEPT2 (SLC15A2) may be a critical factor in the modulation of opioidergic neurotransmission in vivo.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kidney H(+)/peptide cotransporter; Oligopeptide transporter, kidney isoform; Peptide transporter 2; proton-dependent high affinity oligopeptide transporter PepT2; Slc15a2 variant A; Slc15a2 variant A2; solute carrier family 15 (H+/peptide transporter), member 2; solute carrier family 15 (oligopeptide transporter), member 2; Solute carrier family 15 member 2; solute carrier family 15, member 2
Gene Aliases: 8430408C16Rik; C78862; PEPT2; SLC15A2
UniProt ID: (Human) Q16348
Entrez Gene ID: (Human) 6565, (Mouse) 57738, (Rat) 60577
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support