Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Peroxiredoxin (Prx) is a growing peroxidase family, whose mammalian members have been known to connect with cell proliferation, differentiation, and apoptosis. Many isoforms (about 50 proteins), collected in accordance to the amino acid sequence homology, containing active site cysteine residue, and the thiol-specific antioxidant activity, distribute throughout all the kingdoms. Among them, mammalian Prx consists of 6 different members grouped into typical 2-Cys, atypical 2-Cys Prx, and 1-Cys Prx. Except Prx VI belonging to 1-Cys Prx subgroup, the other five 2-Cys Prx isotypes have the thioredoxin-dependent peroxidase (TPx) activity utilizing thioredoxin, thioredoxin reductase, and NADPH as a reducing system. Mammalian Prxs are 20-30 kilodalton in molecular size and vary in subcellular localization: Prx I, II, and VI in cytosol, Prx III in mitochondria, Prx IV in ER and secretion, Prx V showing complicated distribution including peroxisome, mitochondria and cytosol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anti-oxidant protein 1; antioxidant protein; Antioxidant protein 1; AOP-1; HBC189; mitochondrial thioredoxin dependent peroxide reductase; mitochondrial Trx dependent peroxide reductase; Perioredoxin-3; Peroxiredoxin III; Peroxiredoxin-3; Protein MER5; Protein MER5 homolog; Prx 3; PRX III; Prx-III; Thioredoxin-dependent peroxide reductase, mitochondrial; Thioredoxin-dependent peroxiredoxin 3
Gene Aliases: AOP-1; AOP1; AW822249; D0Tohi1; Ef2l; HBC189; MER5; PRDX3; PRO1748; prx-III; Prx3; SP-22; SP22; TDXM
UniProt ID: (Human) P30048, (Mouse) P20108
Entrez Gene ID: (Human) 10935, (Mouse) 11757
Molecular Function: peroxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support