Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
KCTD12 gene ontology annotations related to this gene include protein homooligomerization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain-containing protein KCTD12; Pfetin; potassium channel tetramerisation domain containing 12; Predominantly fetal expressed T1 domain; testicular tissue protein Li 100
Gene Aliases: C13orf2; KCTD12; KIAA1778; PFET1; PFETIN
UniProt ID: (Human) Q96CX2
Entrez Gene ID: (Human) 115207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support