Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Antigen Affinity Purified from Pooled Serum
AKT1 (PKB alpha) is a serine/threonine kinase that regulates cell survival. The activated enzyme inhibits apoptosis and stimulates cell cycle progression by phosphorylating numerous targets in various cell types, including cancer cells. This protein kinase is activated by insulin, PI3K, IGF1 and various other growth and survival factors. Akt promotes cell survival by inhibiting apoptosis through phosphorylation and inactivation of several targets, including forkhead transcription factors, and caspase-9. The AKT pathway is a major target for cancer drug discovery.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Akt kinase; Akt1-PA; Akt1-PB; Akt1-PC; Akt1-PD; Akt1-PE; C-AKT; CAKT; CG4006-PA; CG4006-PB; CG4006-PC; CG4006-PD; CG4006-PE; DAkt; dAkt kinase; PKB; PKBG; Protein kinase B; RAC serine/threonine protein kinase; RAC serine/threonine-protein kinase; related to PKA to PKC protein kinases; related to the A and C kinases; SD10374p
Gene Aliases: AKT; AKT/PKB; AKT1; Akt1|PKB; CG4006; D-Akt; DAKT; dAKT/dPKB; dAkt/PKB; DAKT1; DAKT1/PKB; Dmel\CG4006; Dmel_CG4006; DPKB; DRAC-PK; DRAC-PK66; DRAC-PK85; l(3)04226; l(3)89Bq; P-AKT; pAkt; PKB; PKB/AKT; PKB/dAKT; RAC; RacPK
UniProt ID: (Fruit fly) Q8INB9
Entrez Gene ID: (Fruit fly) 41957
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support