Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a member of the Fe65 protein family. It is an adaptor protein localized in the nucleus. It interacts with the Alzheimer's disease amyloid precursor protein, transcription factor CP2/LSF/LBP1 and the low-density lipoprotein receptor-related protein. APP functions as a cytosolic anchoring site that can prevent the gene product's nuclear translocation. This encoded protein could play an important role in the pathogenesis of Alzheimer's disease. It is thought to regulate transcription. Also it is observed to block cell cycle progression by downregulating thymidylate synthase expression. Multiple alternatively spliced transcript variants have been described for this gene but some of their full length sequence is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adaptor protein FE65a2; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); amyloid beta A4 precursor protein-binding family B member 1; Amyloid beta precursor protein binding family B member 1; Amyloid-beta A4 precursor protein-binding family B member 1; Protein Fe65; stat-like protein
Gene Aliases: APBB1; FE65; MGC:9072; RIR
UniProt ID: (Human) O00213, (Rat) P46933, (Mouse) Q9QXJ1
Entrez Gene ID: (Human) 322, (Rat) 29722, (Mouse) 11785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support