Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Cell treatment to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 4
Predicted band size: 301 kDa
Positive Control: HeLa cell lysate, HeLa treated with Hydroxyurea cell lysate, HeLa treated with 4mM Hydroxyurea for 20 hours.
Subcellular Location: Chromosome, Nucleus.
Sequence Similarities: No Cross with Mouse and Rat.
Tissue Specificity: Ubiquitous, with highest expression in testis. Isoform 2 is found in pancreas, placenta and liver but not in heart, testis and ovary.
The protein encoded by this gene is a serine/threonine kinase and DNA damage sensor, activating cell cycle checkpoint signaling upon DNA stress. The encoded protein can phosphorylate and activate several proteins involved in the inhibition of DNA replication and mitosis, and can promote DNA repair, recombination, and apoptosis. This protein is also important for fragile site stability and centrosome duplication. Defects in this gene are a cause of Seckel syndrome 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2310008J16Rik; 2810405N18Rik; Ataxia telangiectasia and Rad3-related protein; EC 2.7.11.1; FRAP-related protein 1; FRAP-related protein-1; kinase ATR; MEC1, mitosis entry checkpoint 1, homolog; protein kinase ATR; Serine/threonine-protein kinase ATR; TEM8
Gene Aliases: ATR; FCTCS; FRP1; MEC1; SCKL; SCKL1
UniProt ID: (Human) Q59HB2
Entrez Gene ID: (Human) 545
Molecular Function:
non-receptor serine/threonine protein kinase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support