Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody detects a 42 kDa* protein corresponding to the molecular mass of Actin on SDS-PAGE immunoblots of pervanadate treated human C2C12 and SYF cSrc transformed cells, but not in control cells. In addition, this antibody either detects actin cleavage products at 36 and 20 kDa, or cross-reacts with unidentified proteins at these molecular weights in C2C12 cells.
Actin exists as a ubiquitous protein involved with filament formation that make up large portions of the cytoskeleton. Actin filaments interact with myosin to assist in muscle contraction as well as aiding in cell motility and cytokinesis. In vertebrates there are three groups of actin isoforms: alpha, beta and gamma. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exists in most cell types as components of the cytoskeleton and as mediators of internal cell motility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; Actin, alpha cardiac muscle 1; Actin, alpha skeletal muscle; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 2; actin, gamma, cytoplasmic 1; Actin, gamma-enteric smooth muscle; actin-like protein; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; Alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; Atcb; beta cytoskeletal actin; Beta-actin; beta-actin FE-3; Cell growth-inhibiting gene 46 protein; cytoplasmic beta-actin; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; epididymis luminal protein 176; Gamma-2-actin; Gamma-actin; gamma-enteric smooth muscle actin; nemaline myopathy type 3; pan actin; PS1TP5-binding protein 1; RP5-1068B5.2; smooth muscle alpha-actin; Smooth muscle gamma-actin
Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACT; Act-4; Act4; ACTA; Acta-2; ACTA1; ACTA2; ACTA3; ACTB; ACTC; Actc-1; ACTC1; ACTE; ACTG; ACTG1; ACTG2; ACTGE; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; ACTVS; Actx; AL023024; alphaSMA; ASD5; ASMA; beta-actin; BRWS1; BRWS2; CFTD; CFTD1; CFTDM; CMD1R; CMH11; DFNA20; DFNA26; E430023M04Rik; E51; GIG46; HEL-176; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMalphaA; SMGA; VSCM
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support