Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Phospho-BIK (Ser35) Polyclonal Antibody detects endogenous levels of BIK only when phosphorylated at Ser35.
The Bik protein is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programmed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apoptosis inducer NBK; apoptosis-inducing NBK; BBC1; Bcl-2-interacting killer; BCL2-interacting killer (apoptosis-inducing); bhikhari; BIP 1; BIP1; BP 4; BP4; cb 60; natural born killer
Gene Aliases: BIK; BIP1; BP4; NBK
UniProt ID: (Human) Q13323
Entrez Gene ID: (Human) 638
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support