Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: Synthetic peptide: 1:8000
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C/EBP delta are CCAAT enhancer binding proteins (CEBPs), a family of transcription factors that all contain a highly conserved, basic-leucine zipper domain at the C-terminus that is involved in dimerization and DNA binding. At least six members of the family have been isolated and characterized to date (CEBP alpha to CEBP zeta). CEBPD is a leucine zipper (LZ) DNA-binding protein that regulates gene expression in a variety of tissues including liver, adipose, lung and intestine. CEBPD is an important transcriptional activator in the regulation of genes involved in immune and inflammatory responses and has been reported to possess many tumor suppressor-like properties.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C EBP; C/EBP delta; CCAAT/enhancer binding protein (C/EBP), delta; CCAAT/enhancer-binding protein delta; NF-IL6-beta; Nuclear factor NF-IL6-beta
Gene Aliases: C/EBP-delta; CEBPD; CELF; CRP3; NF-IL6-beta
UniProt ID: (Human) P49716
Entrez Gene ID: (Human) 1052
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support