Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
This antibody was cross-adsorbed to phospho-tyrosine coupled to agarose then affinity purified using phospho-Integrin beta4 (Tyr-1494) peptide (without carrier). The antibody detects a 200kDa* protein corresponding to the molecular mass of Integrin beta4 on SDS-PAGE immunoblots of A431 cells treated with pervanadate, but not in control cells. Similar results were also observed in src-transformed Hct116 cells treated with pervanadate.
Integrins are heterodimers comprised of alpha and beta subunits, that are noncovalently associated transmembrane glycoprotein receptors. Different combinations of alpha and beta polypeptides form complexes that vary in their ligand-binding specificities. Integrins mediate cell-matrix or cell-cell adhesion, and transduced signals that regulate gene expression and cell growth. This gene encodes the integrin beta 4 subunit, a receptor for the laminins. This subunit tends to associate with alpha 6 subunit and is likely to play a pivotal role in the biology of invasive carcinoma. Mutations in this gene are associated with epidermolysis bullosa with pyloric atresia. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD antigen CD104; CD104; CD104 antigen; GP150; Integrin beta-4
Gene Aliases: AA407042; C230078O20; CD104; GP150; ITGB4
UniProt ID: (Human) P16144, (Mouse) A2A863, (Rat) Q64632
Entrez Gene ID: (Human) 3691, (Mouse) 192897, (Rat) 25724
Molecular Function: integrin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support