Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD32; CD32 receptor 2; CD32 receptor 2a; CD32 receptor 2b; CD32A; CDw32; Fc fragment of IgG, low affinity II, receptor for (CD32); Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity IIb, receptor for (CD32); Fc gamma receptor IIB; Fc gamma RIIB; Fc receptor, IgG, low affinity IIb; FC-gamma RII; fc-gamma RII-a; Fc-gamma RII-b; Fc-gamma-RII; fc-gamma-RIIa; Fc-gamma-RIIb; FcgammaRIIB; FcGR; FCGR2A1; FcRII; fcRII-a; fcRII-b; FGFR2B; IgG Fc receptor II beta; IgG Fc receptor II-b; Low affinity immunoglobulin gamma Fc region receptor II; Low affinity immunoglobulin gamma Fc region receptor II-b; Ly-17; Lymphocyte antigen 17
Gene Aliases: AI528646; CD32; CD32B; F630109E10Rik; Fc[g]RII; FCG2; FcgammaRIIB2; FCGR2; Fcgr2a; FCGR2B; FcgRII; Fcr-2; Fcr-3; FCRII; IGFR2; Ly-17; Ly-m20; LyM-1
UniProt ID: (Human) P31994, (Rat) Q63203, (Mouse) P08101
Entrez Gene ID: (Human) 2213, (Rat) 289211, (Mouse) 14130
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support