Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of CPI17alpha only when phosphorylated at Threonine 38.
CPI-17 is a phosphorylation-dependent inhibitory protein for smooth muscle myosin phosphate. CPI-17 was originally identified as a PKC-potentiated inhibitory protein of protein phosphatase-1, which is dominantly expressed in smooth muscle. Phosphorylation at Threonine 38, in vitro, by PKC or Rho-kinase enhances the inhibitory potency toward myosin phosphatase. CPI-17 is also phosphorylated at Threonine 38 by protein kinase N and might be involved in the calcium sensitization of smooth muscle contraction as a downstream effector of Rho and/or arachidonic acid. CPI-17 is dually phosphorylated at Serine 12 and Threonine 38 by a MYPT-associated kinase, M110 kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 17 kDa PKC-potentiated inhibitory protein of PP1; 17-kDa PKC-potentiated inhibitory protein of PP1; 17-KDa protein; CPI-17; PKC-potentiated inhibitory protein of PP1; Protein kinase C-potentiated inhibitor protein of 17 kDa; Protein phosphatase 1 regulatory subunit 14A; protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Aliases: 1110001M11Rik; Cpi; CPI-17; CPI17; PPP1INL; PPP1R14A
UniProt ID: (Human) Q96A00, (Rat) Q99MC0, (Mouse) Q91VC7
Entrez Gene ID: (Human) 94274, (Rat) 114004, (Mouse) 68458
Molecular Function: phosphatase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support