Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of CUTL1 only when phosphorylated at Ser1215.
The protein encoded by this gene is a member of the homeodomain family of DNA binding proteins. It may regulate gene expression, morphogenesis, and differentiation and it may also play a role in the cell cycle progession. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCAAT displacement protein; CDP; CDP/Cux p200; CDP2; cut homolog; cut-like 1; cut-like homeobox 1; Cux/CDP homeoprotein; golgi integral membrane protein 6; Homeobox protein cut-like 1; Homeobox protein cux-1; Protein CASP; putative protein product of Nbla10317
Gene Aliases: CASP; CDP; CDP/Cut; CDP1; Clox; COY1; CUTL1; CUX; Cux-1; Cux/CDP; CUX1; GOLIM6; Kiaa4047; Nbla10317; p100; p110; p200; p75
UniProt ID: (Human) Q13948, (Mouse) P53564
Entrez Gene ID: (Human) 1523, (Rat) 116639, (Mouse) 13047
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support