Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
7TM Antibodies
FIGURE: 1 / 1
This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana. The cannabinoid receptors are members of the guanine-nucleotide-binding protein coupled receptor family, which inhibit adenylate cyclase activity in a dose-dependent, stereoselective and pertussis toxin-sensitive manner. The two receptors have been found to be involved in the cannabinoid-induced CNS effects experienced by users of marijuana. Multiple transcript variants encoding two different protein isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CANN6; Cannabinoid receptor 1; cannabinoid receptor 1 (brain); CB-R; CB1 receptor; central cannabinoid receptor; HGNC:2159; OTTHUMP00000016839; RP1-23D17.1
Gene Aliases: CANN6; CB-R; CB1; CB1A; CB1K5; CB1R; CNR; CNR1
UniProt ID: (Human) P21554
Entrez Gene ID: (Human) 1268
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support