Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This antibody is specific for human Cenexin-1 protein phosphorylated at S796. A BLAST analysis was used to suggest cross-reactivity with Cenexin-1 from human, chimpanzee and macaque based on 100% homology with the immunizing sequence. Cross-reactivity with Cenexin-1from other sources has not been determined.
Seems to be a major component of sperm tail outer dense fibers (ODF). ODFs are filamentous structures located on the outside of the axoneme in the midpiece and principal piece of the mammalian sperm tail and may help to maintain the passive elastic structures and elastic recoil of the sperm tail. May have a modulating influence on sperm motility. Functions as a general scaffold protein that is specifically localized at the distal/subdistal appendages of mother centrioles. Component of the centrosome matrix required for the localization of PLK1 and NIN to the centrosomes. Required for the formation and/or maintenance of normal CETN1 assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 134; Cenexin; cenexin 1; Outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; Outer dense fiber protein 2; RP11-187B3.1; sperm tail structural protein; testis tissue sperm-binding protein Li 51e
Gene Aliases: CT134; ODF2; ODF2/1; ODF2/2; ODF84
UniProt ID: (Human) Q5BJF6
Entrez Gene ID: (Human) 4957, (Rhesus monkey) 717505
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support