Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
7TM Antibodies
FIGURE: 1 / 1
Chemokine-like receptor 1 (CMKLR1) is a receptor for the chemoattractant adipokine chemerin/RARRES2 and for the omega-3 fatty acid derived molecule resolvin E1. Interaction with RARRES2 induces activation of intracellular signaling molecules, such as SKY, MAPK1/3 (ERK1/2), MAPK14/P38MAPK and PI3K leading to multifunctional effects, like, reduction of immune responses, enhancing of adipogenesis and angionesis. Resolvin E1 down-regulates cytokine production in macrophages by reducing the activation of MAPK1/3 (ERK1/2) and NF-kappa-B. CMKLR1 positively regulates adipogenesis and adipocyte metabolism and acts as a coreceptor for several SIV strains (SIVMAC316, SIVMAC239, SIVMACL7E-FR and SIVSM62A), as well as a primary HIV-1 strain (92UG024-2).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chemerin receptor; Chemerin-like receptor 1; chemokine receptor-like 1; Chemokine-like receptor 1; G-protein coupled receptor ChemR23; G-protein coupled receptor DEZ; orphan G-protein coupled receptor, Dez; resolvin E1 receptor
Gene Aliases: CHEMERINR; CHEMR23; CMKLR1; DEZ; RVER1
UniProt ID: (Human) Q99788
Entrez Gene ID: (Human) 1240
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support