Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May mediate the transduction of intracellular signals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Crk-like protein; crk-like protein {ECO:0000250|UniProtKB:P46109}; crkl {ECO:0000312|EMBL:AAH85865.1, ECO:0000312|RGD:1308531}; OTTHUMP00000198539; v-crk avian sarcoma virus CT10 oncogene homolog-like; v-crk sarcoma virus CT10 oncogene homolog-like
Gene Aliases: 1110025F07Rik; AA589403; AI325100; CRKL; Crkol
UniProt ID: (Human) P46109, (Mouse) Q8BGC5, (Rat) Q5U2U2
Entrez Gene ID: (Human) 1399, (Mouse) 12929, (Rat) 287942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support