Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-EDC3 pS161 is specific for phospho S161 mouse EDC3. A BLAST analysis was used to suggest cross-reactivity with rabbit, pig, monkey, bovine, rat, mouse, and bat based on 100% sequence homology. Cross-reactivity with EDC3 pS161 from other sources has not been determined.
This gene encodes a protein that is important in mRNA degradation. The encoded protein is a component of a decapping complex that promotes efficient removal of the monomethylguanosine (m7G) cap from mRNAs, as part of the 5' to 3' mRNA decay pathway. Mutations in this gene have been identified in human patients with an autosomal recessive form of intellectual disability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Enhancer of mRNA-decapping protein 3; YjeF domain-containing protein 1
Gene Aliases: AA517853; Edc3; Lsm16; Yjdc
UniProt ID: (Mouse) Q8K2D3
Entrez Gene ID: (Mouse) 353190
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support