Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Antibody detects endogenous levels of eIF4B only when phosphorylated at Ser504.
The eukaryotic translation initiation factor 4B (eIF4B) plays a critical role in recruiting the 40S ribosomal subunit to the mRNA. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-4B; Eukaryotic translation initiation factor 4B; hCG_2014609
Gene Aliases: 2310046H11Rik; AL024095; C85189; EIF-4B; Eif4a2; EIF4B; PRO1843
UniProt ID: (Human) P23588, (Mouse) Q8BGD9
Entrez Gene ID: (Human) 1975, (Rat) 300253, (Mouse) 75705
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support