Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Antibody detects endogenous levels of ENSA only when phosphorylated at Ser67.
The protein encoded by this gene belongs to a highly conserved cAMP-regulated phosphoprotein (ARPP) family. This protein was identified as an endogenous ligand for the sulfonylurea receptor, ABCC8/SUR1. ABCC8 is the regulatory subunit of the ATP-sensitive potassium (KATP) channel, which is located on the plasma membrane of pancreatic beta cells and plays a key role in the control of insulin release from pancreatic beta cells. This protein is thought to be an endogenous regulator of KATP channels. In vitro studies have demonstrated that this protein modulates insulin secretion through the interaction with KATP channel, and this gene has been proposed as a candidate gene for type 2 diabetes. At least eight alternatively spliced transcript variants encoding distinct isoforms have been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-endosulfine; ARPP-19e; endosulfine-alpha variant 1; endosulfine-alpha variant 3
Gene Aliases: 1700020C18Rik; 2610007F17Rik; AI451924; ARPP-19e; ENSA
UniProt ID: (Human) O43768, (Rat) P60841, (Mouse) P60840
Entrez Gene ID: (Human) 2029, (Rat) 60334, (Mouse) 56205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support