Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
These proteins belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene encodes a protein that binds ephrin-A ligands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific kinase; CEK-7; eck-like sequence 1; EHK-1; EK4; EK7; EK8; EPH homology kinase 1; EPH-like kinase 4; EPH-like kinase 7; EPH-like kinase 8; Eph-like tyrosine kinase 1; Ephrin type-A receptor 3; Ephrin type-A receptor 4; Ephrin type-A receptor 5; H TYRO4; HEK; Human embryo kinase; human embryo kinase 1; receptor protein-tyrosine kinase HEK7; receptor protein-tyrosine kinase HEK8; rEK4; testicular tissue protein Li 64; TYRO1 protein tyrosine kinase; TYRO4 protein tyrosine kinase; tyrosine-protein kinase receptor EHK-1; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase receptor MPK-3; tyrosine-protein kinase receptor REK4; Tyrosine-protein kinase receptor SEK; Tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1; Tyrosine-protein kinase TYRO4
Gene Aliases: 2900005C20Rik; AI385584; AI854630; AW125296; AW492086; BSK; Cek4; CEK7; Cek8; EHK-1; EHK1; EK4; EK7; Ekh1; Els1; Els1.; End3; EPHA3; EPHA4; EPHA5; ETK; ETK1; HEK; HEK4; HEK7; HEK8; Mek4; rb; Rek4; Rek7; RGD1560587; SEK; Sek1; TYRO1; TYRO4
UniProt ID: (Human) P29320, (Human) P54764, (Human) P54756, (Rat) O08680, (Mouse) Q03137, (Mouse) Q60629, (Rat) P54757
Entrez Gene ID: (Human) 2042, (Human) 2043, (Human) 2044, (Mouse) 13837, (Rat) 29210, (Rat) 316539, (Mouse) 13838, (Mouse) 13839, (Rat) 79208
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support