Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Antibody detects endogenous levels of EZH2 only when phosphorylated at Thr487.
EZH2 (Histone-lysine N-methyltransferase) is a member of the Polycomb-group (PcG) family. PcG family members form multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes over successive cell generations. This protein associates with the embryonic ectoderm development protein, the VAV1 oncoprotein, and the X-linked nuclear protein. EZH2 may play a role in the hematopoietic and central nervous systems. Mutations affecting EZH2 can cause Weaver syndrome (WVS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: an; Enhancer of zeste homolog 2; ENX 1; ENX-1; EZH 2; EZH-2; Histone-lysine N-methyltransferase EZH2; Lysine N-methyltransferase 6; OTTHUMP00000213774; OTTHUMP00000213778; OTTHUMP00000213779; OTTHUMP00000213780
Gene Aliases: ENX-1; ENX1; Enx1h; EZH1; EZH2; EZH2b; KMT6; KMT6A; mKIAA4065; WVS; WVS2
UniProt ID: (Human) Q15910, (Mouse) Q61188
Entrez Gene ID: (Human) 2146, (Rat) 312299, (Mouse) 14056
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support