Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Eph receptor A2 is the receptor for members of the ephrin-A family. It binds to ephrin-A1, -A3, -A4 and -A5. Eph receptor A3 is also a receptor for members of the ephrin-A family. It binds to ephrin-A2, -A3, -A4 and -A5 and is thought to play a role in lymphoid function. Eph receptor A4 is a receptor for members of the ephrin-A family. It binds to ephrin-A1, -A4 and -A5. It binds more poorly to ephrin-A2 and -A3. It may play a role in a signal transduction process involved in hindbrain pattern formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EK4; EPH-like kinase 4; Eph-like tyrosine kinase 1; ephrin receptor EphA2; Ephrin type-A receptor 2; Ephrin type-A receptor 3; Epithelial cell kinase; epithelial cell receptor protein tyrosine kinase; HEK; Human embryo kinase; human embryo kinase 1; rEK4; soluble EPHA2 variant 1; testicular tissue protein Li 64; TYRO4 protein tyrosine kinase; Tyrosine-protein kinase receptor ECK; Tyrosine-protein kinase receptor ETK1; Tyrosine-protein kinase receptor MPK-5; tyrosine-protein kinase receptor REK4; Tyrosine-protein kinase receptor SEK-2; Tyrosine-protein kinase TYRO4
Gene Aliases: ARCC2; AW492086; AW545284; Cek4; CTPA; CTPP1; CTRCT6; ECK; EK4; End3; EPHA2; EPHA3; ETK; ETK1; HEK; HEK4; Mek4; Myk2; Rek4; Sek-2; Sek2; TYRO4
UniProt ID: (Human) P29317, (Human) P29320, (Mouse) Q03145, (Rat) O08680
Entrez Gene ID: (Human) 1969, (Human) 2042, (Mouse) 13836, (Rat) 366492, (Mouse) 13837, (Rat) 29210
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support