Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Acts as a guanine nucleotide exchange factor (GEF) which differentially activates the GTPases RHOA, RAC1 and CDC42. Plays a role in axon guidance regulating ephrin-induced growth cone collapse and dendritic spine morphogenesis. Upon activation by ephrin through EPHA4, the GEF activity switches toward RHOA resulting in its activation. Activated RHOA promotes cone retraction at the expense of RAC1- and CDC42-stimulated growth cone extension.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain-enriched SH3-domain protein Besh3; Eph-interacting exchange protein; Ephexin-1; Neuronal guanine nucleotide exchange factor; transforming immortalised mammary epithelial sequence 2
Gene Aliases: ARHGEF27; Besh3; EPHEXIN; NGEF; Tims2
UniProt ID: (Human) Q8N5V2, (Mouse) Q8CHT1
Entrez Gene ID: (Human) 25791, (Mouse) 53972, (Rat) 246217
Molecular Function: guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support