Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Phospho-FGFR Pan (Tyr653, Tyr654) Polyclonal Antibody detects endogenous levels of FGFR Pan only when phosphorylated at Tyr653, Tyr654.
This gene encodes a protein that is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds both acidic and basic fibroblast growth factors and is involved in limb induction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bacteria-expressed kinase; Basic fibroblast growth factor receptor 1; BEK fibroblast growth factor receptor; bFGF-R; bFGF-R-1; BFGFR; c-fgr; CD331; CD332; CD333; CD334; CTLA-2-beta protein; FGF receptor-1; fgfr 1; FGFR-1; FGFR-2; FGFR-3; FGFR-4; FGFR1/PLAG1 fusion; Fibroblast growth factor receptor 1; Fibroblast growth factor receptor 2; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; Fibroblast growth factor receptor 4; fibroblast growth factor receptor 4 16 minus form; FLT-2; FMS-like tyrosine kinase 2; fms-related tyrosine kinase 2; heparin-binding growth factor receptor; hydroxyaryl-protein kinase; K-sam; Keratinocyte growth factor receptor; KGFR; MFR; N-sam; protein tyrosine kinase, receptor like 14; protein-tyrosine kinase; Protein-tyrosine kinase receptor MPK-11; Proto-oncogene c-Fgr; tyrosine kinase JTK4; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase
Gene Aliases: ACH; AU043015; AW208770; AW556123; BBDS; BEK; bFGF-R-1; BFGFR; BFR-1; c-fgr; CD331; CD332; CD333; CD334; CEK; CEK2; CEK3; CFD1; Eask; ECCL; ECT1; FGFBR; FGFR-1; Fgfr-2; Fgfr-3; Fgfr-4; Fgfr-7; FGFR1; FGFR2; FGFR3; FGFR4; Fgfr7; FLG; Flg-2; FLT-2; FLT2; FR3; HBGFR; HH2; HRTFDS; HSFGFR3EX; Hspy; JTK2; JTK4; JWS; K-SAM; KAL2; KGFR; KGFRTr; KSAM; MFR; Mfr3; Mpk-11; N-SAM; OGD; sam3; svs; TK14; TK25; TKF
UniProt ID: (Human) P11362, (Human) P21802, (Human) P22607, (Human) P22455, (Rat) Q04589, (Mouse) P16092, (Mouse) P21803, (Rat) Q498D6, (Mouse) Q03142
Entrez Gene ID: (Human) 2260, (Human) 2263, (Human) 2261, (Human) 2264, (Rat) 79114, (Mouse) 14182, (Mouse) 14183, (Rat) 25022, (Rat) 84489, (Mouse) 14184, (Rat) 25114, (Mouse) 14186
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support